1H-Indazole-3-carboxaldehyde 98%
Proveedor: BLD PHARMATECH GMBH
|
|
BLDPBD58789-100GEA
1360
EUR
BLDPBD58789-100G
BLDPBD58789-10G
BLDPBD58789-250MG
BLDPBD58789-25G
BLDPBD58789-1G
BLDPBD58789-5G
1H-Indazole-3-carboxaldehyde 98%
1H-Indazole-3-carboxaldehyde
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₈H₆N₂O MW: 146,146 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD04972468 Núm. CAS: 5235-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...