1H-Imidazo[4,5-b]pyridine-2-thiol 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD58739-250MGEA 35 EUR
BLDPBD58739-250MG BLDPBD58739-1G BLDPBD58739-100MG BLDPBD58739-5G BLDPBD58739-25G
1H-Imidazo[4,5-b]pyridine-2-thiol 95%
1H-Imidazo[4,5-b]pyridine-2-thiol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₅N₃S
MW: 151,189 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00022663
Núm. CAS: 29448-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR