2-Nitrophenylacetylene 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD58335-10GEA 414 EUR
BLDPBD58335-10G BLDPBD58335-100MG BLDPBD58335-25G BLDPBD58335-5G BLDPBD58335-250MG BLDPBD58335-1G
2-Nitrophenylacetylene 97%
2-Nitrophenylacetylene

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₅NO₂
MW: 147,131 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD01113917
Núm. CAS: 16433-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR