4-Bromo-3-(trifluoromethyl)anisole 98%

BLDPBD5827-100GEA 239 EUR
BLDPBD5827-100G BLDPBD5827-1G BLDPBD5827-25G BLDPBD5827-10G BLDPBD5827-5G
4-Bromo-3-(trifluoromethyl)anisole 98%
4-Bromo-3-(trifluoromethyl)anisole

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆BrF₃O
MW: 255,04 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08059527
Núm. CAS: 400-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR