3-Chloro-N,N-dimethylbenzamide 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD582554-10GEA 151 EUR
BLDPBD582554-10G BLDPBD582554-5G BLDPBD582554-250MG BLDPBD582554-1G
3-Chloro-N,N-dimethylbenzamide 98%
3-Chloro-N,N-dimethylbenzamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₀ClNO
MW: 183,64 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01214035
Núm. CAS: 24167-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR