(2-Bromo-5-chlorophenyl)acetic acid 97%

BLDPBD5820-500GEA 939 EUR
BLDPBD5820-500G BLDPBD5820-1G BLDPBD5820-5G BLDPBD5820-100G BLDPBD5820-10G BLDPBD5820-25G BLDPBD5820-250MG
(2-Bromo-5-chlorophenyl)acetic acid 97%
(2-Bromo-5-chlorophenyl)acetic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆BrClO₂
MW: 249,4900 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00153890
Núm. CAS: 81682-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR