1-Bromo-2-(methoxymethyl)benzene 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD57526-250MGEA 10 EUR
BLDPBD57526-250MG BLDPBD57526-5G BLDPBD57526-1G BLDPBD57526-25G
1-Bromo-2-(methoxymethyl)benzene 98%
1-Bromo-2-(methoxymethyl)benzene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₉BrO
MW: 201,0605 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03790887
Núm. CAS: 52711-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR