Ciclohex-1-enilmetilcetona 95%

BLDPBD56170-1GEA 34 EUR
BLDPBD56170-1G BLDPBD56170-25G BLDPBD56170-5G
Ciclohex-1-enilmetilcetona 95%
Ciclohex-1-enilmetilcetona
Formula: C₈H₁₂O
MW: 124,18 g/mol
Número MDL: MFCD00001547
Núm. CAS: 932-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR