10,12-Tricosadiynoic acid 98%

BLDPBD55681-250MGEA 62 EUR
BLDPBD55681-250MG BLDPBD55681-100MG
10,12-Tricosadiynoic acid 98%
10,12-Tricosadiynoic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₂₃H₃₈O₂
MW: 346,56 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00041683
Núm. CAS: 66990-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR