1,4-Thioxane 1,1-dioxide 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD54811-1GEA 10 EUR
BLDPBD54811-1G BLDPBD54811-5G
1,4-Thioxane 1,1-dioxide 98%
1,4-Thioxane 1,1-dioxide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄H₈O₃S
MW: 136,16 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011683
Núm. CAS: 107-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR