1,4-Dipropylbenzene 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD54770-250MGEA 25 EUR
BLDPBD54770-250MG
1,4-Dipropylbenzene 97%
1,4-Dipropylbenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₈
MW: 162,2713 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03844789
Núm. CAS: 4815-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR