Ethyl-2-piperidinecarboxylate 98%

BLDPBD5465-10GEA 13 EUR
BLDPBD5465-10G BLDPBD5465-5G BLDPBD5465-25G
Ethyl-2-piperidinecarboxylate 98%
Ethyl-2-piperidinecarboxylate

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₈H₁₅NO₂
MW: 157,21 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00005980
Núm. CAS: 15862-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR