1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD53660-1GEA 18 EUR
BLDPBD53660-1G BLDPBD53660-25G BLDPBD53660-250MG BLDPBD53660-10G BLDPBD53660-5G BLDPBD53660-100G
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester 95%
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₂H₂₁BN₂O₂
MW: 236,12 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD06659062
Núm. CAS: 844891-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR