1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride 97%

BLDPBD53657-5GEA 182 EUR
BLDPBD53657-5G BLDPBD53657-250MG BLDPBD53657-100MG BLDPBD53657-1G
1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride 97%
1,3,5-Trimethyl-1H-pyrazole-4-sulphonylchloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₉ClN₂O₂S
MW: 208,6700 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD02180944
Núm. CAS: 59340-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR