1,10-Phenanthroline hydrochloride 99%
|
|
BLDPBD52226-25GEA
17
EUR
BLDPBD52226-25G
1,10-Phenanthroline hydrochloride 99%
1,10-Phenanthroline hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₂H₉ClN₂ MW: 216,66 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00150061 Núm. CAS: 3829-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...