2'-Fluoro-5'-nitroacetophenone 98%

BLDPBD50062-1GEA 6 EUR
BLDPBD50062-1G BLDPBD50062-100G BLDPBD50062-0.025G BLDPBD50062-10G BLDPBD50062-500G BLDPBD50062-25G BLDPBD50062-5G
2'-Fluoro-5'-nitroacetophenone 98%
2'-Fluoro-5'-nitroacetophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆FNO₃
MW: 183,1400 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00277867
Núm. CAS: 79110-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR