1-(2-Aminoethyl)piperidine 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD49822-25GEA 98 EUR
BLDPBD49822-25G BLDPBD49822-5G BLDPBD49822-1G
1-(2-Aminoethyl)piperidine 98%
1-(2-Aminoethyl)piperidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₁₆N₂
MW: 128,22 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00006516
Núm. CAS: 27578-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR