3-Chloroisoquinoline-5-boronic acid 90%

BLDPBD493349-250MGEA 239 EUR
BLDPBD493349-250MG BLDPBD493349-1G BLDPBD493349-100MG
3-Chloroisoquinoline-5-boronic acid 90%
3-Chloroisoquinoline-5-boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₇BClNO₂
MW: 207,421 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD22199341
Núm. CAS: 1429665-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR