4-Cyanophenylhydrazine hydrochloride 98%
|
|
BLDPBD4807-100GEA
103
EUR
BLDPBD4807-100G
BLDPBD4807-5G
BLDPBD4807-10G
BLDPBD4807-1G
BLDPBD4807-25G
BLDPBD4807-500G
4-Cyanophenylhydrazine hydrochloride 98%
4-Cyanophenylhydrazine hydrochloride
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₇H₈ClN₃ MW: 169,61 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00673994 Núm. CAS: 2863-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...