4-Chloro-6-nitro-o-toluidine 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD4770-5GEA 61 EUR
BLDPBD4770-5G BLDPBD4770-0.025G BLDPBD4770-1G BLDPBD4770-25G
4-Chloro-6-nitro-o-toluidine 98%
4-Chloro-6-nitro-o-toluidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₇ClN₂O₂
MW: 186,60 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01320687
Núm. CAS: 62790-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR