(Methoxycarbonylmethyl)triphenylphosphonium bromide 97%

BLDPBD46481-5GEA 6 EUR
BLDPBD46481-5G BLDPBD46481-500G BLDPBD46481-1000G BLDPBD46481-100G BLDPBD46481-25G BLDPBD46481-10G
(Methoxycarbonylmethyl)triphenylphosphonium bromide 97%
(Methoxycarbonylmethyl)triphenylphosphonium bromide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₁H₂₀BrO₂P
MW: 415,28 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011801
Núm. CAS: 1779-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR