3-Boronobenzenesulphonamide 98%

BLDPBD44986-250MGEA 149 EUR
BLDPBD44986-250MG BLDPBD44986-100MG BLDPBD44986-25MG BLDPBD44986-1G
3-Boronobenzenesulphonamide 98%
3-Boronobenzenesulphonamide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₈BNO₄S
MW: 201,00 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06659840
Núm. CAS: 850568-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR