2-Fluoro-5-formylphenylboronic acid 98%

BLDPBD4477-100GEA 630 EUR
BLDPBD4477-100G BLDPBD4477-1G BLDPBD4477-5G BLDPBD4477-25G
2-Fluoro-5-formylphenylboronic acid 98%
2-Fluoro-5-formylphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₆BFO₃
MW: 167,93 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03095100
Núm. CAS: 352534-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR