(S)-1-Methyl-3-(hydroxymethyl)piperidine 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD447655-250MGEA 73 EUR
BLDPBD447655-250MG BLDPBD447655-1G BLDPBD447655-5G
(S)-1-Methyl-3-(hydroxymethyl)piperidine 98%
(S)-1-Methyl-3-(hydroxymethyl)piperidine

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₁₅NO
MW: 129,2 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11035736
Núm. CAS: 205194-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR