2-Nitro-4-thiocyanatoaniline 97%

BLDPBD4445-250MGEA 6 EUR
BLDPBD4445-250MG BLDPBD4445-10G BLDPBD4445-25G BLDPBD4445-100G BLDPBD4445-5G BLDPBD4445-1G
2-Nitro-4-thiocyanatoaniline 97%
2-Nitro-4-thiocyanatoaniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₅N₃O₂S
MW: 195,2000 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00463878
Núm. CAS: 54029-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR