5-Iodosalicylaldehyde 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD4433-5GEA 49 EUR
BLDPBD4433-5G BLDPBD4433-100G BLDPBD4433-250MG BLDPBD4433-10G BLDPBD4433-1G BLDPBD4433-25G
5-Iodosalicylaldehyde 98%
5-Iodosalicylaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₅IO₂
MW: 248,0200 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00723501
Núm. CAS: 1761-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR