Iminostilbene 97%

BLDPBD4419-500GEA 306 EUR
BLDPBD4419-500G BLDPBD4419-5G BLDPBD4419-25G BLDPBD4419-100G
Iminostilbene 97%
Iminostilbene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₄H₁₁N
MW: 193,25 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00005071
Núm. CAS: 256-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR