1,4-Difluoro-2-methyl-5-nitrobenzene 97%

BLDPBD4405-100GEA 225 EUR
BLDPBD4405-100G BLDPBD4405-25G BLDPBD4405-250MG BLDPBD4405-10G BLDPBD4405-5G BLDPBD4405-1G
1,4-Difluoro-2-methyl-5-nitrobenzene 97%
1,4-Difluoro-2-methyl-5-nitrobenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₅F₂NO₂
MW: 173,1169 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06200905
Núm. CAS: 141412-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR