4-Hydroxy-3-nitrobenzonitrile 98%

BLDPBD4261-25GEA 45 EUR
BLDPBD4261-25G BLDPBD4261-10G BLDPBD4261-5G
4-Hydroxy-3-nitrobenzonitrile 98%
4-Hydroxy-3-nitrobenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₄N₂O₃
MW: 164,12 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00070775
Núm. CAS: 3272-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR