5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4-disulphonic acid monosodium salt 98% (contains < 2% H₂O)
Proveedor: BLD PHARMATECH GMBH
|
|
BLDPBD41961-250MGEA
32
EUR
BLDPBD41961-250MG
BLDPBD41961-5G
BLDPBD41961-1G
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4-disulphonic acid monosodium salt 98% (contains < 2% H₂O)
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4-disulphonic acid monosodium salt
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₂₀H₁₃N₄NaO₆S₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00078417 Núm. CAS: 69898-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...