Thieno[3,2-b]thiophene 98%

BLDPBD41503-250MGEA 7 EUR
BLDPBD41503-250MG BLDPBD41503-1G BLDPBD41503-25G BLDPBD41503-10G BLDPBD41503-100G BLDPBD41503-5G
Thieno[3,2-b]thiophene 98%
Thieno[3,2-b]thiophene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₄S₂
MW: 140,23 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00179479
Núm. CAS: 251-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR