3,4-Difluoro-o-toluic acid 97%

BLDPBD41329-250MGEA 7 EUR
BLDPBD41329-250MG BLDPBD41329-5G BLDPBD41329-25G BLDPBD41329-1G BLDPBD41329-10G BLDPBD41329-100G
3,4-Difluoro-o-toluic acid 97%
3,4-Difluoro-o-toluic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆F₂O₂
MW: 172,13 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07777153
Núm. CAS: 157652-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR