3-Bromo-5-chlorobenzaldehyde 98%

BLDPBD4120-100GEA 863 EUR
BLDPBD4120-100G BLDPBD4120-1G BLDPBD4120-5G BLDPBD4120-25G BLDPBD4120-250MG BLDPBD4120-10G
3-Bromo-5-chlorobenzaldehyde 98%
3-Bromo-5-chlorobenzaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₄BrClO
MW: 219,4600 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06797226
Núm. CAS: 188813-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR