Benzyltrimethylammonium tribromide 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD40793-1GEA 6 EUR
BLDPBD40793-1G BLDPBD40793-0.5G BLDPBD40793-100G BLDPBD40793-25G BLDPBD40793-5G BLDPBD40793-500G BLDPBD40793-10G
Benzyltrimethylammonium tribromide 97%
Benzyltrimethylammonium tribromide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₁₆Br₃N
MW: 389,97 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00134423
Núm. CAS: 111865-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR