4-Cyano-2-fluorophenylboronic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD38047-250MGEA 66 EUR
BLDPBD38047-250MG BLDPBD38047-5G BLDPBD38047-1G
4-Cyano-2-fluorophenylboronic acid 98%
4-Cyano-2-fluorophenylboronic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₅BFNO₂
MW: 164,93 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD07644472
Núm. CAS: 1150114-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR