Ethyl-2-amino-4-methylthiazole-5-carboxylate 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD3802-1GEA 6 EUR
BLDPBD3802-1G BLDPBD3802-100G BLDPBD3802-10G BLDPBD3802-5G BLDPBD3802-25G
Ethyl-2-amino-4-methylthiazole-5-carboxylate 97%
Ethyl-2-amino-4-methylthiazole-5-carboxylate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₁₀N₂O₂S
MW: 186,22 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00123414
Núm. CAS: 7210-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR