3-Amino-2,6-dimethylpyridine 98%

BLDPBD3778-1GEA 7 EUR
BLDPBD3778-1G BLDPBD3778-10G BLDPBD3778-5G BLDPBD3778-100G BLDPBD3778-25G
3-Amino-2,6-dimethylpyridine 98%
3-Amino-2,6-dimethylpyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₁₀N₂
MW: 122,17 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01646409
Núm. CAS: 3430-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR