Tiofeno-2-etilamina 98%

BLDPBD3761-25GEA 15 EUR
BLDPBD3761-25G BLDPBD3761-100G BLDPBD3761-10G BLDPBD3761-5G BLDPBD3761-1000G BLDPBD3761-500G
Tiofeno-2-etilamina 98%
Tiofeno-2-etilamina

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₉NS
MW: 127,21 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00051495
Núm. CAS: 30433-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR