2,4-Dimethoxy-5-pyrimidineboronic acid 97%

BLDPBD3728-5GEA 57 EUR
BLDPBD3728-5G BLDPBD3728-500G BLDPBD3728-10G BLDPBD3728-250MG BLDPBD3728-100G BLDPBD3728-25G BLDPBD3728-1G
2,4-Dimethoxy-5-pyrimidineboronic acid 97%
2,4-Dimethoxy-5-pyrimidineboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₆H₉BN₂O₄
MW: 183,9577 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD01318985
Núm. CAS: 89641-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR