3-Bromo-5-phenylsalicylic acid 98%

BLDPBD370457-100MGEA 209 EUR
BLDPBD370457-100MG BLDPBD370457-50MG BLDPBD370457-250MG BLDPBD370457-25MG
3-Bromo-5-phenylsalicylic acid 98%
3-Bromo-5-phenylsalicylic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₃H₉BrO₃
MW: 293,1128 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD18378258
Núm. CAS: 4906-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR