3-Aminophenylboronic acid hydrochloride 97%
|
|
BLDPBD3672-1GEA
11
EUR
BLDPBD3672-1G
BLDPBD3672-5G
BLDPBD3672-25G
3-Aminophenylboronic acid hydrochloride 97%
3-Aminophenylboronic acid hydrochloride
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₆H₉BClNO₂ MW: 173,41 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00191748 Núm. CAS: 85006-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...