Ethyl thiazole-4-carboxylate 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD3654-10GEA 57 EUR
BLDPBD3654-10G BLDPBD3654-25G BLDPBD3654-5G BLDPBD3654-1G
Ethyl thiazole-4-carboxylate 97%
Ethyl thiazole-4-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₆H₇NO₂S
MW: 157,1900 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03788566
Núm. CAS: 14527-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR