Ethyl-4-amino-3-chlorobenzoate 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD35029-5GEA 96 EUR
BLDPBD35029-5G BLDPBD35029-1G BLDPBD35029-250MG
Ethyl-4-amino-3-chlorobenzoate 97%
Ethyl-4-amino-3-chlorobenzoate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₁₀ClNO₂
MW: 199,6300 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06411007
Núm. CAS: 82765-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR