7-Bromo-4-hydroxyquinoline-3-carboxylic acid 97%

BLDPBD35000-10GEA 403 EUR
BLDPBD35000-10G BLDPBD35000-5G BLDPBD35000-1G
7-Bromo-4-hydroxyquinoline-3-carboxylic acid 97%
7-Bromo-4-hydroxyquinoline-3-carboxylic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₀H₆BrNO₃
MW: 268,064 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD02684222
Núm. CAS: 860205-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR