(S)-(+)-2-Amino-1,1-diphenyl-1-propanol 97%

BLDPBD34946-10GEA 297 EUR
BLDPBD34946-10G BLDPBD34946-1G BLDPBD34946-250MG BLDPBD34946-5G
(S)-(+)-2-Amino-1,1-diphenyl-1-propanol 97%
(S)-(+)-2-Amino-1,1-diphenyl-1-propanol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₅H₁₇NO
MW: 227,302 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00145253
Núm. CAS: 78603-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR