2-(Trimethylsilyl)ethoxymethyl chloride 90% estabilizado
|
|
Estabilización:
Stabilised with diisopropylethylamine
BLDPBD34874-25GEA
46
EUR
BLDPBD34874-25G
BLDPBD34874-0.025G
2-(Trimethylsilyl)ethoxymethyl chloride 90% estabilizado
2-(Trimethylsilyl)ethoxymethyl chloride
Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C
Formula:
C₆H₁₅ClOSi MW: 166,72 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00009919 Núm. CAS: 76513-69-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...