2,2,2,3'-Tetrafluoroacetophenone 97%

BLDPBD34701-250MGEA 7 EUR
BLDPBD34701-250MG BLDPBD34701-1G BLDPBD34701-5G BLDPBD34701-25G
2,2,2,3'-Tetrafluoroacetophenone 97%
2,2,2,3'-Tetrafluoroacetophenone

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₄F₄O
MW: 192,11 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00039236
Núm. CAS: 708-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR