(S)-(-)-Dimetilmalato 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD34323-10GEA 9 EUR
BLDPBD34323-10G BLDPBD34323-5G BLDPBD34323-25G
(S)-(-)-Dimetilmalato 98%
(S)-(-)-Dimetilmalato

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₁₀O₅
MW: 162,1406 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00066215
Núm. CAS: 617-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR