Picolinamidine hydrochloride 97%

BLDPBD33877-1GEA 7 EUR
BLDPBD33877-1G BLDPBD33877-0.025G BLDPBD33877-25G BLDPBD33877-100G BLDPBD33877-5G BLDPBD33877-10G BLDPBD33877-500G
Picolinamidine hydrochloride 97%
Picolinamidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₈ClN₃
MW: 157,60 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00052271
Núm. CAS: 51285-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR