Methyl-5-formylsalicylate 98%

BLDPBD33638-1GEA 7 EUR
BLDPBD33638-1G BLDPBD33638-100G BLDPBD33638-25G BLDPBD33638-5G
Methyl-5-formylsalicylate 98%
Methyl-5-formylsalicylate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₈O₄
MW: 180,1574 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03840784
Núm. CAS: 41489-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR